WypAll™ L20 General Clean™ Medium Duty Wipes BL 7317, Extra Long Jumbo Roll
WypAll™ L20 General Clean™ Medium Duty Wipes BL 7317, Extra Long Jumbo Roll
Actual product may vary
WypAll™ L20 General Clean™ Medium Duty Wipes BL 7317, Extra Long Jumbo Roll
WypAll™ L20 General Clean™ Medium Duty Wipes BL 7317, Extra Long Jumbo Roll
WypAll™ L20 General Clean™ Medium Duty Wipes BL 7317, Extra Long Jumbo Roll
WypAll™ L20 General Clean™ Medium Duty Wipes BL 7317, Extra Long Jumbo Roll
WypAll™ L20 General Clean™ Medium Duty Wipes BL 7317, Extra Long Jumbo Roll
WypAll™ L20 General Clean™ Medium Duty Wipes BL 7317, Extra Long Jumbo Roll
Blue, 2 ply, single use wiping paper designed for a variety of cleaning and maintenance tasks in process environments. Unique Airflex™ technology delivers exceptional strength and absorbency. Compatible with Kimberly-Clark Professional™ dispensers 6146, 6154 and 6155. Quantity 1 roll x 1,000 sheets.
Have Questions?
Catalog NumberDimensions (L x W)
TYL731739.5 X 23.5 cm
Catalog number TYL7317
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Dimensions (L x W):
39.5 X 23.5 cm
  • Each individual roll contains 1,000 blue, 2 ply sheets
  • Single-use wiping paper designed for a variety of cleaning and maintenance tasks in process environments
  • Unique Airflex™ technology delivers exceptional strength and absorbency
  • Jumbo Roll Format
  • Compatible with Kimberly-Clark Professional™ Dispensers (product codes 6146, 6154 and 6155)
Specifications
Product TypeMedium-duty Industrial Wiper
Length (English)14.56 in.
Length (Metric)37.5 cm
Width (English)14.56 in
Width (Metric)37.5 cm
MaterialPaper
Basis Weight4.197 kg
ColorBlue
Dimensions (L x W)39.5 X 23.5 cm
FormatLarge Roll
Packaging Quantity1000 Pk.
Ply2-ply