Check Order Status
ADP Platelet Aggregation Reagent
Actual product may vary
ADP Platelet Aggregation Reagent
ADP is a lyophilized preparation of adenosine-5-diphosphate and used for the evaluation of qualitative platelet function disorders.
Have Questions?
Catalog Number
Quantity
TYL5366
2 x 1 mL
Catalog number
TYL5366
Price (MYR)
956.00
EA
Add to cart
Quantity:
2 x 1 mL
Price (MYR)
956.00
EA
Add to cart
ADP is a lyophilized preparation of adenosine-5-diphosphate and used for the evaluation of qualitative platelet function disorders.
Reagents are optimized for use with AggRAM, but may be used on any turbidometric aggregometer of similar quality
ADP is a lyophilized preparation of adenosine-5-diphosphate
ADP is for use in routine platelet aggregation studies for the evaluation of qualitative platelet function disorders
The concentration of the reconstituted ADP reagent is 200 μM.
Specifications
Concentration
200 μm
Detectable Analytes
Platelets
For Use With (Equipment)
AggRAM
Sample Type
Plasma
Quantity
2 x 1 mL
Form
Lyophilized
Unit Size
EA
Contents & Storage
2°C to 8°C