Glass pH Electrode
Glass pH Electrode
Actual product may vary
Glass pH Electrode
Have Questions?
Change viewbuttonViewtableView
Catalog NumberpH RangeReference ElectrolyteTemperature (Metric) Operating
TYL52080 to 14CRISOLYT0°C to 80°C
TYL52070 to 14CRYSOLYT A0°C to 80°C
Catalog number TYL5208
Price (MYR)
956.00
EA
Add to cart
pH Range:
0 to 14
Reference Electrolyte:
CRISOLYT
Temperature (Metric) Operating:
0°C to 80°C
Price (MYR)
956.00
EA
Add to cart
Specifications
Connector TypesS7
MaterialGlass
pH Range0 to 14
Temperature (Metric) Operating0°C to 80°C
TypepH Electrode
For Use With (Application)In biology and clinical.
Reference ElectrolyteCRISOLYT
Temperature (Metric) Range0°C to 80°C
Diameter (Metric)3 mm
Unit SizeEA