Check Order Status
Costar™ Transtar™-96 96-Well Liquid Transfer System Accessories: Reservoir Liners
Actual product may vary
Costar™ Transtar™-96 96-Well Liquid Transfer System Accessories: Reservoir Liners
For use with 96-well plates
Have Questions?
Catalog Number
Type
TYL4876
Open Reservoir Liner
Catalog number
TYL4876
Price (MYR)
849.00
EA
Add to cart
Type:
Open Reservoir Liner
Price (MYR)
849.00
EA
Add to cart
For use with 96-well plates
These are sterile replacement open reservoir liners for the Transtar-96™ Well Liquid Transfer system.
Specifications
Material
Plastic
Sterility
Sterile
Type
Open Reservoir Liner
For Use With (Application)
Ideal for repetitively filling multi-channel pipettors
No. per Case
100/Cs.
No. per Pack
1/Pk.
Channels
96