Eco Parking Sign No Parking Anytime Left Arrow
Eco Parking Sign No Parking Anytime Left Arrow
Actual product may vary
Eco Parking Sign No Parking Anytime Left Arrow
ZING Enterprises Eco Safety Sign is pre-drilled for easy mounting. Available in two grades of reflective sheeting.
Have Questions?
Catalog NumberGrade
TYL2277Engineer Grade Prismatic
Catalog number TYL2277
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Grade:
Engineer Grade Prismatic
3M Engineer Grade Prismatic sheeting
  • Recommended for commercial signs and non-critical traffic signs, such as parking and way-finding signs
  • Meets ASTM Type I specifications
  • Outdoor life expectancy of 5-7 years.
3M High Intensity Prismatic sheeting
  • Highly reflective, and recommended for use in traffic, road, construction, and regulatory signs
  • Meets ASTM Type III and ASTM Type IV specifications
  • Complies with the FHWA Minimum Retroreflectivity standard
  • Outdoor life expectancy is 10 years.
The sign is compliant with MUTCD standards. All Zing signs are made in the USA from recycled materials, and support sustainability initiatives and LEED credits.
Specifications
GradeEngineer Grade Prismatic
MaterialRecycled Aluminum
Quantity1 Ea.
Mounting TypeCorner Holes
For Use With (Application)Communication
Height (English)18 in.
Width (English)12 in.
LanguagesEnglish
Dimensions (L x W)18 x 12 in.
Displayed TextNo Parking Anytime Left Arrow
ColorRed on White
Unit SizeEA