Hygienic Wall Bracket, Single Hook Module
Hygienic Wall Bracket, Single Hook Module
Actual product may vary
Hygienic Wall Bracket, Single Hook Module
This single hook module is designed for storing cleaning tools that have a hanging hole. The hook is slid onto the supplied single-base/spacer from either side.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
TYL10122Green
KLC10121Pink
KLC10122Green
KLC10123Blue
KLC10124Red
Catalog number TYL10122
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Green
  • Single hook module can hold products weighing up to 3 kg
  • Easy to dissemble foeaning or replacement
  • FDA compliant raw material
Specifications
Product TypeWall Bracket
ColorGreen
MaterialPolypropylene
Height (English)1.9 in.
Length (English)1.6 in.
Weight (English)0.06 lb.
Width (English)3.1 in.
Unit SizeEA