Stainless Steel Container for Autoclaves
Stainless Steel Container for Autoclaves
Actual product may vary
Stainless Steel Container for Autoclaves
Suitable for media.
Have Questions?
Catalog NumberFor Use With (Equipment)
TTLBSK411-00062840 Bench Autoclave
Catalog number TTLBSK411-0006
Price (MYR)
6,119.00
EA
Add to cart
For Use With (Equipment):
2840 Bench Autoclave
Price (MYR)
6,119.00
EA
Add to cart
For over 90 years Tuttnauer, as a family owned business, has been an industry leader satisfying customer expectations with top quality, high performance products and a dedicated service support team. Tuttnauer's sterilization & infection control products are trusted at over 350,000 installations worldwide including Laboratories, Pharmaceutical Facilities, Hospitals and Clinics.
  • Stainless Steel Container for either bench or vertical autoclave
  • Suitable for media
Specifications
For Use With (Application)Liquid Sterilization, Pipette and Glass Sterilization, Instrument Sterilization (Wrapped or Unwrapped), Biohazard and Waste Sterilisation, Agar Preparation, Specialized Customized Cycles.
For Use With (Equipment)2840 Bench Autoclave
Height (Metric) Exterior160 mm
Length (Metric)400 mm
ModelFront-loading
Unit SizeEA