ELV-D Vertical Autoclave
ELV-D Vertical Autoclave
Actual product may vary
ELV-D Vertical Autoclave
The ELV-D Vertical Autoclave has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.
Have Questions?
Catalog NumberCapacity (Metric)Features
TTL5075ELVP-WR-230160 LSingle Phase, Water Reservoir
Catalog number TTL5075ELVP-WR-230
Price (MYR)
-
Capacity (Metric):
160 L
Features:
Single Phase, Water Reservoir
Designed for your laboratory applications. The 5075ELV-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, Fo control, and more.
  • Advanced Control System - Take Advantage of Tuttnauer's state-of-the-art control system with multi-color display
  • Safe - Automatic pressure locking system. Door will not open when the chamber is overpressured
  • Optional Internal Printing - Documents date, time, temperature and pressure
  • Intuitive Multi-Color Display - Multi-color display for easier reading gives quick access to important information. Color is used to indicate the stage of the cycle
  • High Quality Stainless Steel - 316L grade stainless steel chamber and door with electro-polish finish
  • Diverse Load Types - The 5075ELV-D autoclave comes with 8 factory set cycle programs that cover the processing needs of most laboratories and research centers
Specifications
Capacity (English)35.2 gal.
Capacity (Metric)160 L
Depth (Metric) Chamber75 cm
Diameter (English) Chamber20 in.
Depth (English) Chamber30 in.
Diameter (Metric) Chamber50 cm
Length (English)30.8 in.
Length (Metric)77 cm
Model5075ELV-D
Product TypeAutoclave, Top-loading
Temperature (English)221°F to 280°F
Temperature (Metric)105°C to 138°C
FeaturesSingle Phase, Water Reservoir