Staining Jar with Two Lids
Staining Jar with Two Lids
Actual product may vary
Staining Jar with Two Lids
Staining jar with two specialized lids for both storage and convenient use.
Have Questions?
Catalog NumberMaterial
TSS35-3PMP
Catalog number TSS35-3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Material:
PMP
Staining jar with two specialized lids for both storage and convenient use.
Staining jar supplied with two lids.
  • One lid for when rack is not in use, providing evaporation-free storage of staining liquid
  • Second lid with special opening to allow handle of staining rack to be inserted
  • For use with SIE354
Caution: Xylene can cause etching after two days at room temperature.
Specifications
ColorClear
MaterialPMP
ShapeRectangular
For Use With (Application)Microscopy and Microbiology Staining
Unit SizeEA