TSE Series Stackable Undercounter Laboratory Refrigerators and Freezers
TSE Series Stackable Undercounter Laboratory Refrigerators and Freezers
Actual product may vary
TSE Series Stackable Undercounter Laboratory Refrigerators and Freezers
Space-saving refrigerator and freezer storage for routine lab use. Undercounter models with digital control and temperature alarms can be paired using a convenient stacking kit.Thermo Scientific TSE Series Stackable Combo refrigerator/freezer provide dependable performance for everyday lab use.
Have Questions?
Catalog NumberDoor Style
TSE08CGVGlass (Refrigerator), Solid (Freezer)
Catalog number TSE08CGV
HSN 84184090
Price (MYR)
32,381.00
Each
Add to cart
Door Style:
Glass (Refrigerator), Solid (Freezer)
Price (MYR)
32,381.00
Each
Add to cart
Thermo Scientific TSE Series Stackable Undercounter Laboratory Refrigerators and Freezers provide dependable temperature performance and straightforward operation for routine lab use. Designed for space-saving installation, these units pair easily with a stacking kit to support everyday workflows. Each model includes essential features and a two-year warranty, and all use SNAP and F-Gas compliant hydrocarbon refrigerants for efficient, reliable performance.
  • Designed for everyday research and academic lab applications with good performance and easy-to-use digital controls
  • Quiet operation, less than 53 dB
  • SNAP compliant green refrigerants
  • Auto defrost
  • Four 2 in, casters, front casters are lockable
  • Bright LED interior lighting on glass door models
  • Hi/Lo Temperature, and door ajar failure alarms
  • Rear access port
  • Keyed door locks, compatible with popular E-lock adapters
Stacking Kit accessory required
Specifications
Capacity (English) Freezer3 cu.ft
Capacity (English) Refrigerator3 cu.ft
Capacity (Metric) Freezer77 L
Capacity (Metric) Refrigerator77 L
Temperature Range (Freezer)-15°C to -25°C
Temperature Range (Refrigerator)1°C to 10°C
Temperature (Setpoint)5°C (Refrigerator), -20°C (Freezer)
Door StyleGlass (Refrigerator), Solid (Freezer)
No. of Doors2
Door MaterialGlass
CompressorHermetic
DefrostAutomatic
Monitoring OptionsAlarm
Access SecurityKey Lock
RefrigerantR600a
Cabinet MaterialScratch-resistant Painted Steel
Exterior FinishWhite-coated Steel
Certifications/ComplianceCE
StyleStackable Combo
DisplayDigital
Ports2 rear access port (1 per undercounter)
Warranty2 Years Parts & Labor
Dimensions (DxWxH) Exterior25.3 x 23.4 x 32.8 in. (64.5 x 59.5 x 83.3 cm) x 2
Dimensions (D x W x H) Interior19.3 x 19.0 x 24.4 in. (49.0 x 48.3 x 62 cm) x 2
Plug TypeEU
Voltage230 V
Frequency50 Hz
Unit SizeEach