Methoxamine (MOX) Reagent
Product Image
Methoxamine (MOX) Reagent
Prepare oximes of steroids and ketoacids prior to silylation with the Thermo Scientific™ Methoxamine (MOX) Reagent.
Have Questions?
Catalog NumberFor Use With
TPCTS-45950Preparing Oximes of Steroids and Ketoacids
Catalog number TPCTS-45950
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With:
Preparing Oximes of Steroids and Ketoacids
Prepare oximes of steroids and ketoacids prior to silylation with the Thermo Scientific™ Methoxamine (MOX) Reagent. It prevents the formation of multiple derivatives when enols are present during silylation, thereby ensuring optimal quantitation. The reagent is a 2% solution of methoxyamine–hydrogen chloride in pyridine.
  • 2% methoxyamine·HCl (M.W. 83.51) in pyridine
  • Prevents formation of multiple derivatives when enols are present during silylation
  • Supplied in amber Thermo Scientific Hypo-Vial Sample Storage Vial with septum and crimp top
For Research Use Only. Not for use in diagnostic procedures.
General References:
  1. Chiabrando, C., et al. (1987). Anal. Biochem. 163, 255-262.
  2. Horning, M.G., et al. (1968). Anal. Biochem. 22, 284.
Specifications
For Use WithPreparing Oximes of Steroids and Ketoacids
Standard Concentration95 - 98, 1 - 3%
HazardsCorrosive, Flammable
Item DescriptionHypo-Vial Container; 10 mL
Storage2°C to 8°C
Standard ComponentsPyridine, Methoxyammonium chloride
Unit SizeEach