Search Thermo Fisher Scientific
| Catalog Number | Top Plate Material | Size | Electrical Requirements |
|---|---|---|---|
| SP135930-33Q | Ceramic | 10.25 x 10.25 in. | 220/240 V 50/60 Hz |
Perform life science and biotechnology applications with the help of Thermo Scientific™ Super-Nuova™ Multi-Position Digital Stirring Hotplates. These hotplates offer four independently-controlled stirring positions on one heated, ceramic top. Product includes 4-function memory and a low-temperature adjustment ideal for life science and biotechnology applications. These stirrers are designed for labs with multiple users and require high-volume processing with unmatched performance, safety and controls.
Operating Features
Safety Features
Includes: