Accessories for MaxQ™ 5000 Shakers
Accessories for MaxQ™ 5000 Shakers
Actual product may vary
Accessories for MaxQ™ 5000 Shakers
Increase the utility of the Thermo Scientific™ MaxQ™ 5000 Shaker with the MaxQ 5000 Gassing Manifold and Stainless-Steel Shelf.
Have Questions?
Catalog NumberDescriptionFor Use With (Equipment)
SHK5000-8GMGassing manifold with 8 outletsMaxQ 5000
Catalog number SHK5000-8GM
Price (MYR)
5,089.00
EA
Add to cart
Description:
Gassing manifold with 8 outlets
For Use With (Equipment):
MaxQ 5000
Price (MYR)
5,089.00
EA
Add to cart
Increase the utility of the Thermo Scientific™ MaxQ™ 5000 Shaker with the MaxQ 5000 Gassing Manifold and Stainless-Steel Shelf.
Gassing Manifold
  • Enables direct gassing into flasks for anaerobic cultures
  • Offered with 8 or 16 outlets
  • Easy-access drain system offers convenience in draining and disinfecting
Stainless-Steel Shelf
  • For back wall of chamber
  • L × W: 33.25 × 4.0 in. (84.5 × 10.2cm)
  • Retractable foot casters allow easy manipulation in lab
Specifications
Product TypeGassing Manifold
DescriptionGassing manifold with 8 outlets
For Use With (Equipment)MaxQ 5000
Unit SizeEA