GC Capillary Column Connectors: Replacement Ferrules
GC Capillary Column Connectors: Replacement Ferrules
Actual product may vary
GC Capillary Column Connectors: Replacement Ferrules
Replacement SilTite ferrules for SilTiteμ-Union
Have Questions?
Catalog NumberFor Use With (Equipment)
SGE0735680.1 to 0.25 mm I.D., 0.53 mm I.D.
Catalog number SGE073568
Price (MYR)
1,033.00
PK
Add to cart
For Use With (Equipment):
0.1 to 0.25 mm I.D., 0.53 mm I.D.
Price (MYR)
1,033.00
PK
Add to cart
  • SilTite μ-Union gives low thermal mass - 9mm in length and <0.5g mass
  • Zero dead volume for optimized peak shape
  • Chemically deactivated stainless steel ferrules give system inertness
  • Robust and stable for both high temperature and pressure cycling
Specifications
TypeReplacement μ-Union Ferrule
MaterialStainless Steel
For Use With (Equipment)0.1 to 0.25 mm I.D., 0.53 mm I.D.
Unit SizePK