Replacement SilTite™ Mini Union Ferrules
Product Image
Replacement SilTite™ Mini Union Ferrules
SilTite Mini Union delivers an easy, leak-free installation for capillary columns without the use of any tools
Have Questions?
Catalog NumberDiameter (Metric) Inside
SGE0734720.45 to 0.53 mm
Catalog number SGE073472
Price (MYR)
526.00
PK
Add to cart
Diameter (Metric) Inside:
0.45 to 0.53 mm
Price (MYR)
526.00
PK
Add to cart
  • Specifically designed for connecting capillary columns
  • Connects guard columns to capillary columns
  • Inert and precise glass lined bore
  • Low dead volume
Specifications
MaterialSilTite
TypeMini Union Ferrules
Quantity10/Pk.
Diameter (Metric) Inside0.45 to 0.53 mm
Unit SizePK