Check Order Status
BP624 GC Capillary Columns
Actual product may vary
BP624 GC Capillary Columns
SGE BP624 GC Capillary Columns - A polar phase designed for EPA volatile organics analysis
Have Questions?
Catalog Number
Length (Metric)
Diameter (Metric) Inner
Film Thickness
SGE054836
30 mm
0.53 mm
3 mm
Catalog number
SGE054836
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
30 mm
Diameter (Metric) Inner:
0.53 mm
Film Thickness:
3 mm
Polar phase
EPA organics analysis (EPA 624, 502.2, SW-846, 8240/8260)
Ideal for environmental analyses, volatile organics, alcohol analysis
Specifications
Type
BP624 GC Capillary Column
Diameter (Metric) Inner
0.53 mm
Max. Temperature
230/240°C
Temperature Range
0°C to 240°C
Film Thickness
3 mm
Length (Metric)
30 mm
Polarity
High polar
Quantity
1 Each