SolGel-WAX™ GC Capillary Columns
SolGel-WAX™ GC Capillary Columns
Actual product may vary
SolGel-WAX™ GC Capillary Columns
Comprised of Polyethylene Glycol in a Sol-Gel matarix. Trajan Scientific and Medical™ SolGel-WAX™ is a robust, high temperature polar WAX phase recommended fro highly active compounds.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric) InnerFilm ThicknessParticle Size
SGE05479730 m0.32 mm0.5 μm0.5 μm
SGE05479630 m0.25 mm0.25 μm0.25 μm
Catalog number SGE054797
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
30 m
Diameter (Metric) Inner:
0.32 mm
Film Thickness:
0.5 μm
Particle Size:
0.5 μm
  • Bonded polyethylene glycol encapsulated in synthetic glass (SolGel).
  • Ideal for mixtures containing alcohols, esters, aldehydes, ketones, or aromatics.
  • SolGel technology facilitates interactions between the sample and the phase, reduces column bleed and improves thermal stability.
  • Extremely stable and inert.
  • Columns are individually tested to ensure quality.
  • Replacement for all Wax columns, including acid or base stabilized.
Specifications
Diameter (Metric) Inner0.32 mm
Stationary PhaseSGE™ SolGel-Wax
For Use With (Application)Highly Active Compounds Analysis
Particle Size0.5 μm
Temperature Range30°C to 260/280°C
Film Thickness0.5 μm
Length (Metric)30 m
PolarityPolar
Unit SizeEA