Check Order Status
BPX70 GC Capillary Column
Actual product may vary
BPX70 GC Capillary Column
SGE BPX70 GC Capillary Column is an MS premium, low bleed column with high polarity specifically designed for fatty acid methyl ester (FAME) analysis
Have Questions?
Change view
Catalog Number
Length (Metric)
Diameter (Metric) Inner
SGE054623
60 m
0.25 mm
SGE054622
30 mm
0.25 mm
2 Options
Catalog number
SGE054623
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
60 m
Diameter (Metric) Inner:
0.25 mm
High temperature
Custom designed for separation of FAMEs
Ideal for cis/trans isomer separation
Polar phase
Specifications
Type
GC Capillary Column
Diameter (Metric) Inner
0.25 mm
Stationary Phase
BPX70
Max. Temperature
250/260°C
Temperature Range
50°C to 250/260°C
Film Thickness
0.25 μm
Length (Metric)
60 m