BPX5 Capillary Columns
BPX5 Capillary Columns
Actual product may vary
BPX5 Capillary Columns
High-performance ultralow bleed columns
Have Questions?
Catalog NumberLength (Metric)Temperature Range
SGE05410130 m-40°C to +360/370°C
Catalog number SGE054101
Price (MYR)
4,161.00
EA
Add to cart
Length (Metric):
30 m
Temperature Range:
-40°C to +360/370°C
Price (MYR)
4,161.00
EA
Add to cart
High-performance ultralow bleed columns
  • Nonpolar; extended temperature range
  • Composed of (5%)-phenyl (equiv.) polysilphenylene-siloxane
  • Columns with an inner diameter of 0.25mm are suitable for U.S. EPA Methods 604, 606, 610, 611, 625, 1625, 8040B, 8060, 8090, 8100, 8250, and 8270B
Specifications
Diameter (Metric) Inner0.25 mm
Stationary PhaseSGE™ BPX5
For Use With (Application)Phenols Analysis, Solvents Analysis, Phenols, Amines, Pesticide Analysis, Herbicides Analysis
Particle Size0.25 μm
Temperature Range-40°C to +360/370°C
Film Thickness0.25 μm
Length (Metric)30 m
PolarityNon polar
Unit SizeEA