Check Order Status
Headspace/Soil Gas Syringes
Actual product may vary
Headspace/Soil Gas Syringes
Gives better performance and superior quality. Trajan Scientific and Medical Headspace/Soil Gas Syringes has the knowledge and expertise that goes into manufacturing an SGE syringe.
Have Questions?
Catalog Number
Volume (Metric)
SGE008775
5 mL
Catalog number
SGE008775
Price (MYR)
1,342.00
EA
Add to cart
Volume (Metric):
5 mL
Price (MYR)
1,342.00
EA
Add to cart
High quality solutions for liquid handling
Accurate and reproducible injections
Headspace/soil gas analysis syringes
Specifications
Accuracy
±1%
Description
Headspace Syringe
Diameter (Metric) Inner
0.75 mm
Diameter (Metric) Outer
1.587 mm
Length (Metric)
60 mm
Quantity
1/Pk.
Volume (Metric)
5 mL