Check Order Status
Beckman/Altex, Rheodyne, SSI, Valco Valve Replacement Needle
Actual product may vary
Beckman/Altex, Rheodyne, SSI, Valco Valve Replacement Needle
Removable needle replacement for manual syringes. For use with Beckman/Altex, Rheodyne, SSI, and Valco Valves
Have Questions?
Catalog Number
Volume (Metric)
SGE002313
10 μL
Catalog number
SGE002313
Price (MYR)
437.00
EA
Add to cart
Volume (Metric):
10 μL
Price (MYR)
437.00
EA
Add to cart
High quality solutions for liquid handling
Accurate and reproducible injections
Specifically designed for Beckman/Altex, Rheodyne, Valco, SSI valves
Specifications
Material
PTFE (Plunger)
Description
Gas tight
Needle Gauge
22 G
Diameter (Metric) Inner Needle
0.17 mm
Diameter (Metric) Outer Needle
0.028 mm
Volume (Metric)
10 μL
Needle Point Style
LC