Check Order Status
Sodium Chloride (Crystalline/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisherbrand
Sodium Chloride (Crystalline/Certified ACS), Fisher Chemical™
Have Questions?
Change view
Quantity:
10 kg
Packaging:
Poly Pail
Catalog number
S271-10
Price (MYR)
542.85
Each
Add to cart
Quantity:
10 kg
Packaging:
Poly Pail
Request bulk or custom format
Price (MYR)
542.85
Each
Add to cart
Specifications
Boiling Point
1461°C
Chemical Name or Material
Sodium Chloride
Melting Point
801°C
CAS
7647-14-5
ChemAlert Storage Symbol
Gray
View more