Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Actual product may vary
Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Maximize storage space with these shelf kits for Thermo Scientific™ Ultra-Low Temperature Freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsFor Use With (Equipment)No. of Racks
RSK600SD4Boxes (150/6 Racks)Thermo Scientific TSX and TDE Series (28.8 cu.ft.)6
RSK500SD4Boxes (125/5 Racks)Thermo Scientific TSX and TDE Series (24.1 cu.ft.)5
Catalog number RSK600SD4
Price (MYR)
12,072.00
EA
Add to cart
Holds:
Boxes (150/6 Racks)
For Use With (Equipment):
Thermo Scientific TSX and TDE Series (28.8 cu.ft.)
No. of Racks:
6
Price (MYR)
12,072.00
EA
Add to cart
Maximize storage space with Shelf Kits with Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers, featuring 2 in. boxes with dividers.
Features:
  • Stainless steel racks
  • Boxes
  • Cell dividers
Warranty varies by country. Please contact us for details.
Specifications
Cell Dividers100
Product TypeShelf Kit
DescriptionShelf Kit
HoldsBoxes (150/6 Racks)
MaterialStainless Steel
Shelves4 Tiers
For Use With (Equipment)Thermo Scientific TSX and TDE Series (28.8 cu.ft.)
No. of Racks6
Unit SizeEA