Glove Box 855-AC Anaerobic Chamber
Glove Box 855-AC Anaerobic Chamber
Actual product may vary
Glove Box 855-AC Anaerobic Chamber
Manually control your atmosphere when working with oxygen-sensitive materials in this glove box. Plas Labs™ Glove Box 855-AC Anaerobic Chamber uses a catalyst heater to reduce any trace entering oxygen to water vapor.
Have Questions?
Catalog NumberDepth (Metric)
PLA855-ACEXP
also known as PLA855-AC/EXP
89 cm
Catalog number PLA855-ACEXP
also known as PLA855-AC/EXP
Price (MYR)
-
Depth (Metric):
89 cm
  • Standard 24-month warranty (not applicable for gloves and consumables)
  • Formed one-piece clear acrylic top
  • Self-sealing quick disconnects enable the operator to change the drying train without disturbing the internal atmosphere
  • Matched die-molded thermoset white bottom
  • Adjustable vacuum gauge on transfer chamber
  • Two vacuum pressure pumps, one each for transfer chamber and drying train
  • Four ground key cock valves for purging
  • Drying train includes polycarbonate canisters containing DesiSphere™
  • Inner door on transfer chamber opens automatically when pressures are neutralized between main chamber and transfer chamber
  • All controls are illuminated
  • Transfer chamber is 12in. long and 11in. in diameter (I.D.)
  • Bright Light system (40,000 hour lamp guarantee)
  • Electrical outlet strip; U.L. & C.S.A. approved, hospital grade with four outlets
  • White ambidextrous Hypalon gloves
  • All clamps are adjustable to compensate for wear
Specifications
Product TypeControlled Atmosphere Chamber
Depth (Metric)89 cm
Depth (Metric) Interior71 cm
Height (Metric)97 cm
Height (Metric) Interior66 cm
Volume (Metric)489 L
Weight (English)250 lb.
Width (Metric)140 cm
Width (Metric) Interior104 cm