Color Coded Multi-Purpose Labeling Tape, 1 in. (25.4 mm) Roll
Color Coded Multi-Purpose Labeling Tape, 1 in. (25.4 mm) Roll
Actual product may vary
Color Coded Multi-Purpose Labeling Tape, 1 in. (25.4 mm) Roll
One-sided tape used primarily in laboratories and similar environments for labeling purposes. The 0.50 x 500 in. (12.7 mm x 12.7 m) roll is available in a variety of colors, allowing it to be used as part of a color-coding system. Designed to be removable and versatile for various labeling needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
OPTST-103Green
OPTST-106Blue
Catalog number OPTST-103
Price (MYR)
1,081.00
EA
Add to cart
Color:
Green
Price (MYR)
1,081.00
EA
Add to cart
  • Resistant to water, oil and acid
  • Temperature range of -20 to +121°C
  • For autoclave, freezer, refrigerator and oven
  • Universal marking with ballpoint pen, paper pencil and felt marker
Specifications
AutoclavableAutoclavable
Adhesive TypeRemovable
ColorGreen
FormatRoll
Product TypeLabel Tape
For Use With (Application)Ideal for Universal Marking Pen, a Pencil and Felt
For Use With (Equipment)For Autoclave, Freezer, Fridge and Oven
Quantity1 Ea.
Temperature Range (Metric)-20°C to +121°C
Dimensions (LxW)500 x 1 in.
Length (English)500 in.
Length (Metric)12.7 m
Width (English)1 in.
Width (Metric)25.4 mm
Diameter (English)1 in.
Unit SizeEA