Starter Water Analysis Portable pH Meters
Starter Water Analysis Portable pH Meters
Actual product may vary
Starter Water Analysis Portable pH Meters
LCD meters combine portability with precise pH measurement.
Have Questions?
Catalog NumberModel
OHA83033961ST300
Catalog number OHA83033961
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Model:
ST300
  • Auto buffer recognition stores calibration data to avoid errors
  • Automatic/manual temperature compensation for accurate readings
  • ORP or temperature electrode connectivity for additional functionality
  • IP54 protection shields water/dust damage
  • Size: 90 x 150 x 35 mm
  • Weight: 0.18 kg
Specifications
Accuracy (Temperature)±0.5°C
Accuracy (mV)±1
Accuracy (pH)±0.01
Data Management Details30 Sets, most recent calibration data
Includes1 Plastic gel electrode, buffer powder sachet
Measurement ModespH with mV and Temperature
CalibrationAutomatic/Manual
Range (pH)0 to 14
Resolution (pH)0.01
Range (mV)-1999 to 1999
Resolution (mV)1 mV
Calibration Points1 to 3
Temperature Range (Metric)0°C to 100°C
Resolution (Temperature)0.1°C
Display TypeLCD
Backlight OptionNone
Data Points30
Log Function TypeAutomatic Data Logging
InputsBNC, NTC
IP RatingIP54
Weight (Metric)0.18 kg
MaterialABS
ModelST300
Resolution0.01 pH; 1 mV; 0.1°C
Battery TypeAAA
TypePortable pH Meters
Battery Life>500 hr.
Width (English)3.5 in.
Width (Metric)9 cm
Height (English)1.3 in.
Height (Metric)3.5 cm
Depth (Metric)17 cm
Unit SizeEA