CD79B Antibody (iladatuzumAb) - Humanized, Novus Biologicals™
CD79B Antibody (iladatuzumAb) - Humanized, Novus Biologicals™
Actual product may vary
CD79B Antibody (iladatuzumAb) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Have Questions?
Catalog NumberQuantity
OHA3004564150 μg
Catalog number OHA30045641
Price (MYR)
3,350.00
EA
Add to cart
Quantity:
50 μg
Price (MYR)
3,350.00
EA
Add to cart
CD79B Monoclonal antibody specifically detects CD79B in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
AntigenCD79B
ApplicationsFlow Cytometry, ELISA, Functional Assay
ClassificationMonoclonal
CloneiladatuzumAb
ConjugateUnconjugated
FormulationLyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Gene Accession No.P40259
Gene AliasB29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein
Host SpeciesHuman
ImmunogenCD79b
Purification MethodProtein A purified
Quantity50 μg
Regulatory StatusRUO
Research DisciplineImmunology
Primary or SecondaryPrimary
Gene ID (Entrez)974
ReconstitutionReconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Target SpeciesHuman
FormPurified
IsotypeIgG1
Unit SizeEA
Contents & Storage
Store at -20°C in powder form. Store at -80°C once reconstituted.