11mm Assembled Autosampler Clear Glass Vial Kit, silver seal, PTFE/red rubber septum
Product Image
11mm Assembled Autosampler Clear Glass Vial Kit, silver seal, PTFE/red rubber septum
Save time and reduce the risk of contamination using this convenience kit. It includes 100 clear vials pre-assembled with 100 screw thread caps and septa.
Have Questions?
Catalog NumberColor
NSCC4011-1CVClear
Catalog number NSCC4011-1CV
Price (MYR)
-
Color:
Clear
Save time during sample preparation and reduce the risk of contamination. Each Thermo Scientific™ National™ 11mm Clear Vial Convenience kits includes 100 clear glass vials pre-assembled with 100 screw thread caps and septa. They are packaged in convenient vial trays with clear covers.
  • Aluminum seal is crimped onto the vial following a purge with dry nitrogen
  • Sample is added via injection through the septum
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure ColorSilver
DescriptionClear glass; Silver seal, PTFE/Red rubber septa
ColorClear
MaterialGlass, PTFE, Rubber
Volume (Metric)2 mL
SeptumPTFE/Red rubber
No. per Pack100
Closure MaterialAluminum
Closure TypeCrimp Cap
Closure Size11 mm
Marking SpotNo
Unit SizePK