8mm Autosampler Vial Crimp Caps
8mm Autosampler Vial Crimp Caps
Actual product may vary
8mm Autosampler Vial Crimp Caps
8mm Crimp Top Closures provide a leak-resistant seal, feature 4mm openings and are available with a variety of septa materials with and without liners.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSeptumMaterialType
NSCC4008-1ASilver, Clear, RedClear PTFE/Red RubberPTFE, Rubber, AluminumCrimp Cap
C4008-1ASilver, Clear, RedClear PTFE/Red RubberPTFE, Rubber, AluminumCrimp Cap
Catalog number NSCC4008-1A
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Silver, Clear, Red
Septum:
Clear PTFE/Red Rubber
Material:
PTFE, Rubber, Aluminum
Type:
Crimp Cap
These 8mm Crimp Top Closures feature convenient pre-assembled caps and septa. They provide a secure leak-resistant seal and minimize contamination from handling. 8mm Crimp Top closures feature 4mm openings and are available with a variety of septa materials with and without liners to match your chromatography needs. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
  • Available in a variety of colors and septum types
  • Aluminum seals must be applied with a crimping tool
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionClear PTFE/red rubber septa; Silver
ColorSilver, Clear, Red
MaterialPTFE, Rubber, Aluminum
SeptumClear PTFE/Red Rubber
Quantity200
Thickness (Metric)1.0 mm
TypeCrimp Cap
Closure Size8 mm
For Use With (Equipment)8 mm Crimp Top Vials
Unit SizePK