Check Order Status
OIML Class E1 Weight Set, Aluminum Case
Actual product may vary
OIML Class E1 Weight Set, Aluminum Case
Weight set constructed of polished stainless steel and housed in an aluminum case.
Have Questions?
Catalog Number
Weight (Metric)
NSC303056
1 mg to 500 g
Catalog number
NSC303056
Price (MYR)
8,123.00
EA
Add to cart
Weight (Metric):
1 mg to 500 g
Price (MYR)
8,123.00
EA
Add to cart
Set of weights
Polished stainless steel
Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic case
Specifications
Classification
E1
Material
Stainless Steel
Case Material
Aluminum
Weight (Metric)
1 mg to 500 g