Irradiated Sharpie Fine Point Marker
Irradiated Sharpie Fine Point Marker
Actual product may vary
Irradiated Sharpie Fine Point Marker
Irradiated, fine point permanent marker with fade and water resistant, quick drying, non-toxic ink for use in aseptic environments. Available capped or retractable styles.
Have Questions?
Catalog NumberColorLength (English)Packaging
MICPEN-22IRBlue6 in.Individually bagged, 4 per double bag, 12 per third bag (case pack)
Catalog number MICPEN-22IR
Price (MYR)
433.00
CS
Add to cart
Color:
Blue
Length (English):
6 in.
Packaging:
Individually bagged, 4 per double bag, 12 per third bag (case pack)
Price (MYR)
433.00
CS
Add to cart
Fade and water resistant ink
Micronova Irradiated Sharpie Fine Point Marker is the original pen style, industry standard permanent marker. This cleanroom sharpie marks permanently on most surfaces with quick drying, non-toxic, alcohol based ink that is fade and water resistant to further extend multiple project uses.
  • Fine point sharpie marker
  • Marks permanently on most surfaces
  • Writes on cold or wet surfaces, plastic bags and disposable lab-ware
  • Nontoxic, permanent, quick drying, alcohol-based ink
  • Fade and water resistant
  • Available capped or retractable styles, red, blue or black inks
  • Gamma irradiated for Aseptic ISO 5 and other clean production areas
Specifications
MaterialHDPE
ColorBlue
Cover MaterialHDPE
DescriptionRetractable fine tip permanent marker
Dimensions (L x W x H)6 x 0.5 x 0.5 in. (15.24 x 1.27 x 1.27 cm)
Length (English)6 in.
Length (Metric)15.2 cm
Width (English)0.5 in.
Width (Metric)1.27 cm
AutoclavableNot Autoclavable
Certifications/ComplianceISO 9001 Certified Facility; Certificate of Irradiation Processing; Certificate of Conformance
Diameter (English) Outer0.5 in.
Diameter (Metric) Outer1.27 cm
DisposableReusable
For Use With (Application)Cleanroom and Aseptic Environment Documentation and Labeling
IncludesCap
PackagingIndividually bagged, 4 per double bag, 12 per third bag (case pack)
Product TypeMarker
Quantity12/Pk.
Size8 x 8 x 8 in.
SterilitySterile
Tip Style1.0 mm Fine Point
Unit SizeCS