SlideMop™ Mop Heads, Irradiated
Actual product may vary
SlideMop™ Mop Heads, Irradiated
Mop covers for use with the Micronova™ SlideMop™. The SlideMop is a simple, effective mop system featuring sturdy, collapsible plates that easily slide into the quilted polyester mop cover and lock into place with an effortless downward push.
Have Questions?
Catalog NumberFor Use With (Equipment)
MICMZCR63-16IRStainless Steel Frame
Catalog number MICMZCR63-16IR
Price (MYR)
1,806.00
Case
For Use With (Equipment):
Stainless Steel Frame
Price (MYR)
1,806.00
Case
  • Polyester mop covers for use with the the Micronova SlideMop™ Flat Head Mop. (For SlideMop frame, see QSM16.)
  • Quilted polyester mop covers with polyester pad interiors for disinfectant dispersalManufactured for use in Controlled Environments, ISO 5 thru 9
  • Good chemical resistance
Specifications
Product TypeIrradiated Mop Slipcover
Head TypeFlat
ColorWhite
Length (English)16 in.
SterilitySterile
Length (Metric)41 cm
AutoclavableNot Autoclavable
DisposableReusable
Care InstructionsItem may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C.
For Use With (Equipment)Stainless Steel Frame
For Use With (Application)Cleaning and Sanitization of Cleanroom Surfaces
Color-codedNo
MaterialPolyester
Unit SizeCase