Cartridges for Multidrop™ Pico 1 & Pico 8 Digital Dispensers
Cartridges for Multidrop™ Pico 1 & Pico 8 Digital Dispensers
Actual product may vary
Cartridges for Multidrop™ Pico 1 & Pico 8 Digital Dispensers
The Dispensehead cartidges allow dispensing of one to eight fluids per cassette, from 11 picoliters to 10 microliters of small molecules and biomolecules in DMSO or aqueous solution
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Channels
LTR000411 pL to 20 μL8
LTR00031 nL to 200 μL4
Catalog number LTR0004
Price (MYR)
-
Volume (Metric):
11 pL to 20 μL
Channels:
8
Great choice for dispensing picoliter to microliter volumes for one to eight different types of fluid across an entire plate or just part of a plate.
  • Cost Saving through assay miniaturization
  • Improved efficiency and sample integrity
  • Powerful, flexible, and easy to use software
Specifications
Product TypeDigital Dispenser Cartridge
AutoclavableNot Autoclavable
Channels8
Volume (Metric)11 pL to 20 μL
For Use With (Application)Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting
For Use With (Equipment)Mulitdrop Pico 8 Digital Dispenser
InterfaceUSB
Plate Type12, 24, 48, 96, 384, 1536 well plate, Plate height: 6-47 mm
Precision≤ 8%
SoftwarePicoIT 8
Speed11 pL to 10 μL in seconds
Dimensions (L x W x H)8 x 10.5 x 2.5 cm
Weight (Metric)0.45
Electrical Requirements100 to 240V 50/60Hz
Voltage100/240 V