Check Order Status
Iodine Solution, Wij'S, Pure, Indicator Grade, Fisher Chemical™
Element Search
Structure Search
Iodine Solution, Wij'S, Pure, Indicator Grade, Fisher Chemical™
2.5L, 118 deg.C, Amber, 17 deg.C, 11354, Plastic Coated Glass bottle, Liquid, 2.1, 11mmHg at 20 deg.C
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
LSMJ4500PB17
also known as LSMJ/4500/PB17
Price (MYR)
269.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
269.00
EA
Add to cart
Specifications
Boiling Point
118°C
Melting Point
17°C
CAS
7790-99-0
Color
Amber
pH
2
View more
2.5L, 118 deg.C, Amber, 17 deg.C, 11354, Plastic Coated Glass bottle, Liquid, 2.1, 11mmHg at 20 deg.C