CHECKIT™ Comparator Test Kits: Chlorine-Free/Total
CHECKIT™ Comparator Test Kits: Chlorine-Free/Total
Actual product may vary
CHECKIT™ Comparator Test Kits: Chlorine-Free/Total
Accurate test kits for routine water analysis
Have Questions?
Catalog NumberRange
LOV1470000.02 to 0.3mg/L
Catalog number LOV147000
Price (MYR)
705.00
EA
Add to cart
Range:
0.02 to 0.3mg/L
Price (MYR)
705.00
EA
Add to cart
Accurate test kits for routine water analysis
  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
Specifications
Accuracy±5%
For Use With (Application)Routine water analysis
IncludesAqua comparator™ housing, aqua comparator™ disc D55, DPD No. 1 tablets, DPD No. 3 tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
No. of Tests100
Range0.02 to 0.3mg/L
TypeChlorine Free and Total Test Kit
Unit SizeEA