Clear Stoppering Chamber
Clear Stoppering Chamber
Actual product may vary
Clear Stoppering Chamber
Clear Stoppering Chamber for use with 6 L, 12 L and 18 L Freezone™ Dryers.
Have Questions?
Catalog NumberElectrical Requirements
LBC7868030230 V, 50/60 Hz, International
Catalog number LBC7868030
Price (MYR)
53,651.00
EA
Add to cart
Electrical Requirements:
230 V, 50/60 Hz, International
Price (MYR)
53,651.00
EA
Add to cart
  • Allows flasks attachment with freeze dry system or contain samples in bulk
Specifications
TypeClear Stoppering Chamber
No. of Shelves1
Temperature Range (Metric)Up to 40°C
MaterialAcrylic
For Use With (Application)To provide economical means of stoppering serum bottles
For Use With (Equipment)6 L, 12 L and 18 L FreeZone™ Dryers
Diameter (English)13.5 in.
Diameter (Metric)34 cm
Height (English)23 in.
Height (Metric)58.4 cm
Voltage230 V
Electrical Requirements230 V, 50/60 Hz, International
Frequency50/60 Hz
Weight (English)30 lb.
Weight (Metric)13.6 kg
Unit SizeEA