Check Order Status
Freeze Dry Replacement Parts: Lid Gasket
Actual product may vary
Freeze Dry Replacement Parts: Lid Gasket
Gasket, Lid, Labconco; Replacement, Use on all Labconco Freeze Dry Systems
Have Questions?
Catalog number
LBC7690800
Price (MYR)
-
Gasket, Lid, Labconco; Replacement, Use on all Labconco Freeze Dry Systems
Replacement rubber gasket
Seals chamber lid of FreeZone Freeze Dryer during lyophilization process
Creates tight barrier preventing leakage
Specifications
Type
Lid Gasket
No. of Shelves
0
Material
Rubber
For Use With (Equipment)
For all labconco freeze dry systems