Freeze-Drying Ampule Accessories
Freeze-Drying Ampule Accessories
Actual product may vary
Freeze-Drying Ampule Accessories
Freeze drying ampule accessories allow ampules to be used with Labconco FreeZone Freeze Dryers.
Have Questions?
Catalog NumberType
LBC7576210Stopper
Catalog number LBC7576210
Price (MYR)
351.00
PK
Add to cart
Type:
Stopper
Price (MYR)
351.00
PK
Add to cart
Freeze drying ampule accessories allow ampules to be used with Labconco FreeZone Freeze Dryers.
  • Large rubber split stoppers fit 5, 10, 20, 30, 50, 100 and 125 ml Labconco Serum Bottles (bottles with 20 mm corkage).
    • Package of 100.
  • Oxygen/Natural Gas Sealing Torch is specifically designed for flame sealing freeze dry ampules.
    • Seals all types of heat-resistant glass.
    • Connects to natural gas, butane or propane and oxygen with 1/4 in. ID hose connectors.
  • Ampule valve adapter provides connection of ampules to drying chambers and manifolds by connecting ampules to valve ports or stainless steel stems.
    • Ten per package.
Specifications
TypeStopper
MaterialRubber
Quantity100 Pk.
For Use With (Equipment)Freeze drying system
Diameter (Metric)2 cm
Unit SizePK