Check Order Status
Fast-Freeze™ Flask Filter Paper
Actual product may vary
Fast-Freeze™ Flask Filter Paper
Fits into top of flask to prevent partial loss of product during freeze drying.
Have Questions?
Catalog Number
Diameter (Metric)
LBC7544810
28 mm
Catalog number
LBC7544810
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
28 mm
Fits into top of flask to prevent partial loss of product during freeze drying.
No complex filter retainer needed.
Specifications
Format
Disc
Material
Paper
Pore Size
17 μm
Sterility
Non-sterile
For Use With (Application)
prevents partial loss of product during freeze drying
Quantity
1000/Pk.
Type
Flask Filter
Diameter (English)
1 in.
Diameter (Metric)
28 mm