Drying Chamber Valves for FreeZone™ Freeze-Dry Systems
Drying Chamber Valves for FreeZone™ Freeze-Dry Systems
Actual product may vary
Drying Chamber Valves for FreeZone™ Freeze-Dry Systems
Replacement valves for drying chamber or manifold ports provide leak-resisting high-vacuum seals.
Have Questions?
Catalog number LBC7509600
Price (MYR)
808.00
EA
Add to cart
Price (MYR)
808.00
EA
Add to cart
Replacement valves for drying chamber or manifold ports provide leak-resisting high-vacuum seals.
  • Fabricated of neoprene body and gaskets, molded plastic knob and fittings
  • Valve 10-273-12 accommodates both 1/2 and 3/4 in. adapters
  • Serum bottles fit in valves without adapter
Specifications
TypeDrying Chamber Valve
MaterialNeoprene
For Use With (Application)Drying chambers and manifolds
For Use With (Equipment)Freeze-Dry System
Height (English)3 in.
Height (Metric)7.62 cm
Length (English)2 in.
Weight (English)1 lb.
Weight (Metric)0.5 kg
Unit SizeEA