Shelves for FreeZone™ Freeze Dryers
Shelves for FreeZone™ Freeze Dryers
Actual product may vary
Shelves for FreeZone™ Freeze Dryers
Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeNo. of Shelves
LBC7317100Short3
LBC7317200Tall5
Catalog number LBC7317100
Price (MYR)
8,905.00
EA
Add to cart
Size:
Short
No. of Shelves:
3
Price (MYR)
8,905.00
EA
Add to cart
Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications.
  • Removable, 10 in. diameter aluminum shelves with 1/4 in. rim
  • Shelves slide into stainless steel support
  • Includes 3 rubber feet
  • Design allows for flexible shelf height spacing
  • Shelf can be placed in freezer to prefreeze sample for quick loading
  • Designed to fit inside Drying Chambers (sold separately)
  • Also available: Heated shelves that include ambient to +40°C (+104°F) variable heat control on models manufactured 2016 and earlier and up to +60°C (+140°F) on models 2017 and later (contact your Fisher Customer Service Representative for details).
Specifications
TypeProduct Shelves
Capacity (Metric)150 L
No. of Shelves3
MaterialStainless Steel
SizeShort
For Use With (Application)Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications
For Use With (Equipment)FreeZone™ Drying Chambers
IncludesThree aluminum shelves with 1/4 in. rims, one stainless steel support, and three rubber feet
Width (English)10.4 in.
Height (English)9.8 in.
Height (Metric)24.9 cm
Length (English)10.9 in.
Length (Metric)27.7 cm
Shipping Weight (English)4.5 lb.
Weight (English)3.5 lb.
Unit SizeEA