Refrigerated CentriVap Concentrator Benchtop Vacuum Concentrator with Glass Lid
Refrigerated CentriVap Concentrator Benchtop Vacuum Concentrator with Glass Lid
Actual product may vary
Refrigerated CentriVap Concentrator Benchtop Vacuum Concentrator with Glass Lid
Refrigerated CentriVap Centrifugal Concentrator is designed to rapidly concentrate multiple small heat-sensitive samples.
Have Questions?
Catalog number LBC7310036
Price (MYR)
84,450.00
EA
Add to cart
Price (MYR)
84,450.00
EA
Add to cart
The Refrigerated CentriVap Centrifugal Concentrator is designed to rapidly concentrate multiple small heat-sensitive samples, such as RNA and proteins, using centrifugal force, vacuum and controlled temperature. This exclusive concentrator combines a refrigeration system that cools to -4° C with a heater that warms to 100° C to achieve the user’s desired set point temperatures.
Specifications
CertificationsCE Marked
Shipping Weight (English)108.0 lb.
Shipping Weight (Metric)49.0 kg
Voltage0 to 300 V
Chamber MaterialCorrosion Resistant Steel
Electrical Requirements230 V 50/60 Hz 8 A
Length (English)14.3 in.
Height (English)16.6 in.
Height (Metric)42.2 cm
Length (Metric)36.3 cm
Plug TypeBritish (UK)
Width (Metric)55.1 cm
Width (English)21.7 in.
Dimensions (L x W x H)14.3 x 21.7 x 16.6 in. (36.3 x 55.1 x 42.2 cm)
Unit SizeEA