Check Order Status
Replacement Filters for Benchtop Fume Adsorbers
Actual product may vary
Replacement Filters for Benchtop Fume Adsorbers
Replacement filters for use with Labconco Fume Adsorbers and Fume Adsorber-2s.
Have Questions?
Catalog Number
For Use With (Application)
LBC6961300
Organic Vapor
Catalog number
LBC6961300
Price (MYR)
4,407.00
EA
Add to cart
For Use With (Application):
Organic Vapor
Price (MYR)
4,407.00
EA
Add to cart
Replacement Activated Carbon Filter for use with organic vapors.
6 pounds of activated carbon, adsorbs up to one third of its weight.
Adsorption capacity is dependent on chemical properties.
For use with Labconco Fume Adsorber and Fume Adsorber-2.
Fume Adsorber Models 3955200 and 3955220 include one filter.
Fume Adsorber-2 Models 3955400 and 3955420 include two filters.
Replacement Formaldehyde Carbon Filter for use with formaldehyde and gluteraldehyde.
7.5 pounds of impregnated carbon.
Adsorption weight 10% (0.75 lb.).
Adsorption volume 850 mL of 37% formalin.
For use with Labconco Fume Adsorber and Fume Adsorber-2.
Fume Adsorber Models 3955201 and 3955221 include one filter.
Fume Adsorber-2 Models 3955401 and 3955421 include two filters.
Replacement Ammonia Carbon Filter for use with ammonia and low molecular weight amines.
8.8 pounds of impregnated carbon.
Adsorption weight 10% (0.88 lbs.).
Adsorption volume 800 mL of 50% ammonia.
For use with Labconco Fume Adsorber and Fume Adsorber-2.
Fume Adsorber Models 3955202 and 3955222 include one filter.
Fume Adsorber-2 Models 3955402 and 3955422 include two filters.
Specifications
Filter Type
Carbon
For Use With (Equipment)
Benchtop Fume Adsorber
Weight (English)
11 lb.
Weight (Metric)
5 Kg
Product Type
Filter
For Use With (Application)
Organic Vapor