FlaskScrubber™ Laboratory Glassware Washer Accessory, Glassware Holders
FlaskScrubber™ Laboratory Glassware Washer Accessory, Glassware Holders
Actual product may vary
Labconco
FlaskScrubber™ Laboratory Glassware Washer Accessory, Glassware Holders
For use with FlaskScrubber Laboratory Glassware Washer.
Have Questions?
Catalog NumberMaterialHeight (Metric)
LBC4585204Plastic with Stainless Steel Clip7.62 cm
Catalog number LBC4585204
Price (MYR)
157.00
EA
Add to cart
Material:
Plastic with Stainless Steel Clip
Height (Metric):
7.62 cm
Price (MYR)
157.00
EA
Add to cart
  • Grip glassware over spindles to hold it in place during washing
  • Type 304 stainless steel wire with height adjustment screw
  • May be placed in upper or lower Spindle Rack in Labconco FlaskScrubber
Applications: For flasks or other narrow-neck glassware
Specifications
MaterialPlastic with Stainless Steel Clip
Product TypeGlassware Holder
ColorGray
For Use With (Application)For flasks or other narrow-neck glassware
For Use With (Equipment)FlaskScrubber Laboratory Glassware Washer
Height (English)3 in.
Height (Metric)7.62 cm
Weight (English) Shipping0.4 lb.
Weight (Metric) Shipping181 g
Unit SizeEA