Kimtech™ Pure G3 Cleanroom NXT™ Nitrile Gloves
Actual product may vary
Ansell
Kimtech™ Pure G3 Cleanroom NXT™ Nitrile Gloves
Non-sterile, nitrile gloves do not contain natural rubber latex proteins. Recommended for ISO Class 4 or higher cleanroom environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC62990X-Small
KLC62991Small
KLC62992Medium
KLC62993Large
Catalog number KLC62990
Price (MYR)
1,481.55
Case
Size:
X-Small
Price (MYR)
1,481.55
Case
Non-sterile nitrile gloves do not contain natural rubber latex proteins. Recommended for ISO Class 4 or higher cleanroom environments.
  • Textured fingertips
  • Beaded cuff for easy donning
  • Contain no natural rubber latex
  • Ambidextrous
  • Thickness: 6.2 mil at fingertips
  • Length: 12 in. (30.5 cm)
  • White
  • Meet AQL level of 1.5 for pinholes
  • Packaging: 10 bags of 100
Specifications
Certifications/ComplianceASTM D6978-05: Resistance to permeation by chemo drugs
Cleanroom ClassClass 10 (ISO 4)
ColorWhite
Exterior FinishPartially Textured
Finger Thickness (English)6 mil
Finger Thickness (Metric)0.15 mm
HandAmbidextrous
Latex FreeYes
MaterialNitrile
PowderedPowder Free
SizeX-Small
SterilityNon-sterile
Thickness (English)6 mil
Thickness (Metric)0.15 mm
Model62990
Quantity10 Bags of 100
SeriesKimtech™ G3
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCase