Check Order Status
Filter Disc, Surfaces Plane, Ground Cylindrical, 210 mm
Filter Disc, Surfaces Plane, Ground Cylindrical, 210 mm
The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.
Have Questions?
Change view
Catalog Number
Porosity
KLC62104
4
KLC62101
1
2 Options
Catalog number
KLC62104
Price (MYR)
-
Porosity:
4
Highly resistant to most chemicals and temperatures up to 500°C
The biplane version may be required in specific installation circumstances
Specifications
Material
Borosilicate Glass (3.3)
Sterility
Non-sterile
No. per Pack
1 Ea.
Porosity
4
Thickness (Metric)
11.5 mm ± 0.5 mm
Diameter (Metric)
210 mm
Unit Size
BX