Series 16 4 Porosity Biplane Filter Disc
Product Image
Series 16 4 Porosity Biplane Filter Disc
Have Questions?
Catalog NumberDiameter (Metric)
KLC62004200 mm
Catalog number KLC62004
Price (MYR)
-
Diameter (Metric):
200 mm
Specifications
Quantity1Pack
TypeFilter Disc
Height (Metric)11.5 mm
Diameter (Metric)200 mm
Unit SizeBX