Pure G3 Sterile Latex Gloves
Pure G3 Sterile Latex Gloves
Actual product may vary
Pure G3 Sterile Latex Gloves
Sterile, hand-specific latex gloves are suitable for double-donning and provide sensitive protection. Kimtech™ Pure G3 Sterile Latex Gloves recommended for Class 10/ISO 4 or higher cleanrooms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC568499
KLC568436
KLC568446.5
KLC568457
KLC568467.5
KLC568478
KLC568488.5
KLC5684210
Catalog number KLC56849
Price (MYR)
1,439.00
CS
Add to cart
Size:
9
Price (MYR)
1,439.00
CS
Add to cart
Sterile, hand-specific latex gloves are suitable for double-donning and provide sensitive protection. Recommended for Class 10/ISO 4 or higher cleanrooms.
  • Fully sterilized latex material provides high levels of contamination protection
  • Hand-specific, disposable and a natural color
  • Textured palm and fingertips enhance grip and tactile sensitivity
  • Beaded cuffs add strength to the gloves reducing the risk of tearing
  • Easy donning and doffing
  • Made from latex sterilized with gamma radiation and containing no powder
  • Suitable for use in Class 10/ISO 4 sterile cleanrooms
Specifications
Certifications/ComplianceASTM D3577, ASTM D5151, ASTM D412, ASTM D573, ASTM D3767, ASTM D6319, IEST-RP-CC005, ASTM D5712
Certifications/Compliance (Europe)EN ISO 374-1:2016, EN 374-4:2003, EN ISO 374-5:2016
Cleanroom ClassClass 10 (ISO 4)
Accelerator FreeYes
AQL1.5
ColorNatural
DisposableSingle-use
Exterior FinishFully Textured
Finger Thickness (English)8.7 mil
Finger Thickness (Metric)0.22 mm
HandSpecific
Laboratory/Cleanroom UseControlled Environments
Latex FreeNo
MaterialLatex
Palm Thickness (English)7.9 mil
Palm Thickness (Metric)0.20 mm
PowderedPowder Free
Product TypeCleanroom Gloves
Size9
SterilitySterile
Thickness (English)8 mil
Thickness (Metric)0.20 mm
Model56849
SeriesKimtech™ Pure G3
Tensile Strength24 MPa
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCS