CleanTip™ Swabs: Absorbond™ Series
CleanTip™ Swabs: Absorbond™ Series
Actual product may vary
CleanTip™ Swabs: Absorbond™ Series
No contaminating adhesives
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) TipLength (Metric) Handle
ITWTX7626.6 mm145.2 mm
ITWTX759B3.2 mm59.9 mm
Catalog number ITWTX762
Price (MYR)
-
Diameter (Metric) Tip:
6.6 mm
Length (Metric) Handle:
145.2 mm
No contaminating adhesives
Perfect for abrasion-sensitive applications requiring very low residue such as wiping sensitive-coated optics surfaces.
  • Double-layered, hydroentangled, continuous-filament polyester heads thermally molded onto polypropylene handles without adhesives
  • Ultraclean head materials are precision molded so that every swab is constructed to exacting tolerances
  • Long filaments reduce risk of fiber release; hydroentanglement lowers nonvolatile residue to a very low level
  • Heads are heat molded onto polypropylene handles and perimeter edges heat sealed to prevent fiber release
  • Excellent chemical resistance; can be used with a wide range of solvents
  • Constructed from the cleanest materials available and manufactured in a cleanroom using state-of-the-art processes and equipment
  • High-precision automated process guarantees consistent performance, sizing and cleanliness
  • Silicone-free Bag-Within-A-Bag™ packaging consists of a solvent-resistant, thermally sealed exterior bag that protects the inner bags containing the swabs
  • Each outer bag is individually lot-coded for easy traceability and quality control
Specifications
ConstructionFlexible head paddle, long handle
Diameter (English) Handle0.126 in.
Length (English)5.72 in.
For Use With (Application)Cleaning sensitive surfaces such as optical assemblies, solvent use (such as IPA), cleaning of grooves, tracks, slots and other small spaces
Handle ColorLight Green
Product TypeSwab
Diameter (Metric) Handle3.2 mm
Length (Metric)145.5 mm
Length (English) Handle5.72 in.
Length (Metric) Handle145.2 mm
Thickness (Metric)2.5 mm
Length (English) Tip0.66 in.
Length (Metric) Tip16.7 mm
Width (Metric) Tip3.2 mm
Width (English) Tip0.268 in.
Diameter (Metric) Tip6.6 mm
Diameter (English) Tip0.26 in.
Adhesive TypeThermal
Handle MaterialPolypropylene
Tip MaterialHydroentangled Absorbond™ Polyester
Unit SizeCase