LinerGOLD™ GC Liners
LinerGOLD™ GC Liners
Actual product may vary
LinerGOLD™ GC Liners
Use the gold-standard Thermo Scientific™ LinerGOLD™ GC Liners for consistently high accuracy, sensitivity, and precision in your gas chromatography (GC) analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) InnerLength (Metric)QuantityProduct Type
HYP453T2845-UI2 mm120 mm5PTV Liner, Concentric Baffles
453220442 mm120 mm2PTV Siltek Metal Liner
453500305 mm105 mm5Split Straight Liner
453520572 mm120 mm5PTV Straight Liner
453520621 mm120 mm5PTV Liner, Three Baffles
453T19055 mm105 mm5Split FocusLiner
453T21202 mm120 mm5PTV Baffle Liner (Siltek)
HYP453T21202 mm120 mm5PTV Baffle Liner (Siltek)
453220452 mm120 mm5PTV Glass Straight Liner (Deactivated)
453A1335-UI1 mm78.5 mm5Direct Straight Liner
HYP453A1335-UI1 mm78.5 mm5Direct Straight Liner
Catalog number HYP453T2845-UI
Price (MYR)
2,456.00
PK
Add to cart
Diameter (Metric) Inner:
2 mm
Length (Metric):
120 mm
Quantity:
5
Product Type:
PTV Liner, Concentric Baffles
Price (MYR)
2,456.00
PK
Add to cart
Use the gold standard in GC liner performance and consistency. LinerGOLD GC Liners provide enhanced inertness, leading to increased accuracy, sensitivity, and precision in your gas chromatography (GC) analysis. These injection port liners deliver:
  • Increased accuracy and sensitivity in analysis, preventing the loss of sensitive compounds in analysis
  • Lower detection limits when analyzing active and sensitive compounds
  • Excellent reproducibility gives confidence in your results from liner to liner
  • Consistently high level of performance time after time
LinerGOLD GC liners are manufactured and tested to ensure that you have confidence with every liner that is installed onto your GC system.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Quantity5
Length (Metric)120 mm
Packing MaterialNone
Product TypePTV Liner, Concentric Baffles
Diameter (Metric) Inner2 mm
Diameter (Metric) Outer2.75 mm
For Use With (Equipment)TRACE Ultra and TRACE FOCUS Instruments
Unit SizePK